Potassium nitrite ≥96.0% ACS
Supplier: BeanTown Chemical
143455-100G
143455-25G
143455-500G
BT143455-100GEA
77.93
USD
BT143455-100G
BT143455-25G
BT143455-500G
Potassium nitrite ≥96.0% ACS
Potassium nitrite
CAS: 7758-09-0; EC No: 231-832-4; MDL No: MFCD00011408; RTECS: TT3750000
UN No: UN1488; Haz Class: 5.1; Packing Group: II
Crystalline; Molecular Formula: KNO2; MW: 85.11
Melting Point: 350° (decomposes)
Density (g/mL): 1.915
Hygroscopic
- RTECS: TT3750000
- Hygroscopic
- Crystalline
Formula:
KNO₂ MW: 85.1 g/mol Melting Pt: 350 °C (dec.) Density: 1.915 g/mL |
MDL Number:
MFCD00011408 CAS Number: 7758-09-0 EINECS: 231-832-4 UN: 1488 ADR: 5.1,II |
Specification Test Results
Assay | 96.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...