Magnesium (trace metals basis excluding Ca), powder -100+200 mesh
Supplier: BeanTown Chemical
CAS: 7439-95-4; EC No: 231-104-6; MDL No: MFCD00085308; RTECS: OM2100000
UN No: UN1418; Haz Class: 4.3 (4.2); Packing Group: II
Powder; Molecular Formula: Mg; MW: 24.31
Melting Point: 648°; Boiling Point: 1090°
Density (g/mL): 1.74
- RTECS: OM2100000
- Powder
Formula:
Mg Boiling Pt: 1090 °C Melting Pt: 648 °C Density: 1.74 g/mL |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1418 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...