1,2,4-Trifluorobenzene ≥99%
Supplier: BeanTown Chemical
CAS: 367-23-7; EC No: 206-684-9; MDL No: MFCD00000305
UN No: UN2387; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6H3F3; MW: 132.08
Boiling Point: 88°; Flash point: 4°C (39°F)
Density (g/mL): 1.264; Refractive Index: 1.423
- Refractive index: 1.423
- Liquid
Formula:
C₆H₃F₃ Boiling Pt: 88 °C Density: 1.264 g/mL Flash Pt: 4 °C (39 °F) |
MDL Number:
MFCD00000305 CAS Number: 367-23-7 EINECS: 206-684-9 UN: 2387 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...