5-Bromovaleronitrile ≥98%

Supplier: BeanTown Chemical

143400-25G 143400-5G
BT143400-25GEA 176 USD
BT143400-25G BT143400-5G
5-Bromovaleronitrile ≥98%
5-Bromovaleronitrile

CAS: 5414-21-1; EC No: 226-505-8; MDL No: MFCD00001976 UN No: UN3276; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: Br(CH2)4CN; Molecular Formula: C5H8BrN; MW: 162.03 Density (g/mL): 1.388; Refractive Index: 1.478


  • Refractive index: 1.478
  • Liquid

Formula: BrH₂C(CH₂)₂CH₂CN
Density: 1.388 g/mL
MDL Number: MFCD00001976
CAS Number: 5414-21-1
EINECS: 226-505-8
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR