Sodium chlorate ≥99.0% ACS

Supplier: BeanTown Chemical

143295-100G 143295-2KG 143295-500G
BT143295-100GEA 47.63 USD
BT143295-100G BT143295-2KG BT143295-500G
Sodium chlorate ≥99.0% ACS
Sodium chlorate

CAS: 7775-09-9; EC No: 231-887-4; MDL No: MFCD00003479 UN No: UN1495; Haz Class: 5.1; Packing Group: II Crystalline; Linear Formula: NaClO3; MW: 106.45 Melting Point: 248-261° Density (g/mL): 2.49


  • Crystalline

Formula: ClNaO₃
Melting Pt: 248…261 °C
Density: 2.49 g/mL
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.0% min.
Insoluble matter ≤0.005%
Bromate (BrO3) ≤0.015%
Chloride (Cl) ≤0.005%
Sulfate (SO4) ≤0.001%
Heavy metals (as Pb) ≤0.001%
Iron (Fe) ≤0.0005%
Calcium (Ca) ≤0.002%
Magnesium (Mg) ≤0.002%
Potassium (K) ≤0.01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR