Sodium perchlorate monohydrate 85-90% ACS

Supplier: BeanTown Chemical

143260-1KG 143260-2.5KG 143260-250G
BT143260-1KGEA 154.5 USD
BT143260-1KG BT143260-2.5KG BT143260-250G
Sodium perchlorate monohydrate 85-90% ACS
Sodium perchlorate monohydrate

CAS: 7791-07-3; EC No: 231-511-9; MDL No: MFCD00149164; RTECS: SC9850000 UN No: UN1502; Haz Class: 5.1; Packing Group: II Crystalline; Linear Formula: NaClO4·H2O; MW: 140.48 Melting Point: 130° Density (g/mL): 2.02 Hygroscopic


  • RTECS: SC9850000
  • Hygroscopic
  • Crystalline

Formula: NaClO₄·H₂O
Melting Pt: 130 °C
Density: 2.02 g/mL
MDL Number: MFCD00149164
CAS Number: 7791-07-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II

Order Now

Specification Test Results

Assay 85.0-90.0%
pH of 5% solution at 25°C 6.0-8.0
Insoluble matter ≤0.005%
Chloride (Cl) ≤0.003%
Sulfate (SO4) ≤0.002%
Calcium (Ca) ≤0.02%
Iron (Fe) ≤0.0005%
Heavy metals (as Pb) ≤0.0005%
Potassium (K) ≤0.05%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR