Nickel carbonate, anhydrous ≥98%
Supplier: BeanTown Chemical
Synonyms:
Nickel(II) carbonate, Nickelous Carbonate
143225-250G
143225-50G
BT143225-250GEA
294.72
USD
BT143225-250G
BT143225-50G
Nickel carbonate, anhydrous ≥98%
Nickel carbonate
CAS: 3333-67-3; EC No: 222-068-2; MDL No: MFCD00011144; RTECS: QR6200000
UN No: UN3077; Haz Class: 9; Packing Group: III
-100 Mesh Powder; Linear Formula: NiCO3; MW: 118.70
- RTECS: QR6200000
- -100 Mesh Powder
Formula:
CNiO₃ MW: 118.7 g/mol |
MDL Number:
MFCD00011144 CAS Number: 3333-67-3 EINECS: 222-068-2 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...