Octylamine ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Aminooctane, n-Octylamine, 1-Octylamine

143120-100G 143120-500G
BT143120-100GEA 41.42 USD
BT143120-100G BT143120-500G
Octylamine ≥99%
Octylamine

CAS: 111-86-4; EC No: 203-916-0; MDL No: MFCD00008247; RTECS: RG8050000 UN No: UN2735; Haz Class: 8; Packing Group: II Liquid; Linear Formula: CH3(CH2)7NH2; Molecular Formula: C8H19N ; MW: 129.24 Melting Point: -5°; Boiling Point: 175-177°; Flash point: 60°C (140°F) Density (g/mL): 0.782; Refractive Index: 1.429


  • RTECS: RG8050000
  • Refractive index: 1.429
  • Liquid

Formula: H₃C(CH₂)₆CH₂NH₂
MW: 129.25 g/mol
Boiling Pt: 175…177 °C
Melting Pt: –5 °C
Density: 0.782 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00008247
CAS Number: 111-86-4
EINECS: 203-916-0
UN: 2735
ADR: 8,I

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR