3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-Bromo-5-(trifluoromethyl)benzene-1-sulphonyl chloride, 3-Bromo-5-(trifluoromethyl)benzenesulphonylchloride
143090-1G
143090-5G
BT143090-1GEA
69
USD
BT143090-1G
BT143090-5G
3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride ≥97%
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride
CAS: 351003-46-8; EC No: 000-000-0; MDL No: MFCD03094393
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C7H3BrClF3O2S; MW: 323.51
Boiling Point: 217-218°; Flash point: 65°C (149°F)
Density (g/mL): 1.835; Refractive Index: 1.524
Moisture Sensitive
- Refractive index: 1.524
- Moisture Sensitive
- Liquid
Formula:
C₇H₃BrClF₃O₂S MW: 323.52 g/mol Boiling Pt: 217…218 °C Density: 1.835 g/mL Flash Pt: 65 °C (149 °F) |
MDL Number:
MFCD03094393 CAS Number: 351003-46-8 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...