Indomethacin ≥98%
Supplier: BeanTown Chemical
143065-100G
143065-25G
143065-5G
BT143065-100GEA
658.15
USD
BT143065-100G
BT143065-25G
BT143065-5G
Indomethacin ≥98%
Indomethacin
CAS: 53-86-1; EC No: 200-186-5; MDL No: MFCD00057095
UN No: UN2811; Haz Class: 6.1; Packing Group: I
Solid; Molecular Formula: C19H16ClNO4; MW: 357.80
Melting Point: 158-161°
- Solid
Formula:
C₁₉H₁₆ClNO₄ MW: 357.80 g/mol Melting Pt: 158…161 °C |
MDL Number:
MFCD00057095 CAS Number: 53-86-1 EINECS: 200-186-5 UN: 2811 ADR: 6.1,I |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...