Cobalt selenide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Cobalt(II) selenide
143055-250G
143055-50G
BT143055-250GEA
825
USD
BT143055-250G
BT143055-50G
Cobalt selenide ≥99%
Cobalt selenide
CAS: 1307-99-9; EC No: 215-155-1; MDL No: MFCD00016020
UN No: UN3283; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: CoSe; MW: 137.89
- Powder
Formula:
CoSe MW: 137.89 Melting Pt: 1055 Density: 7.65 |
MDL Number:
MFCD00016020 CAS Number: 1307-99-9 EINECS: 215-155-1 UN: 3283 ADR: 6.1 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...