Tributyltin chloride ≥96%
Supplier: BeanTown Chemical
Synonyms:
Chlorotributylstannane, Tri-n-butyltin chloride, TBTC
143045-1KG
143045-250G
143045-50G
BT143045-1KGEA
262
USD
BT143045-1KG
BT143045-250G
BT143045-50G
Tributyltin chloride ≥96%
Tributyltin chloride
CAS: 1461-22-9; EC No: 215-958-7; MDL No: MFCD00000521; RTECS: WH6820000
UN No: UN2788; Haz Class: 6.1; Packing Group: II
Liquid; Linear Formula: [CH3(CH2)3]3SnCl; Molecular Formula: C12H27ClSn; MW: 325.51
Melting Point: -19°; Boiling Point: 171-173°/25 mmHg; Flash point:
- RTECS: WH6820000
- Refractive index: 1.492
- Liquid
Formula:
C₁₂H₂₇ClSn MW: 325.51 g/mol Boiling Pt: 171…173 °C/25 mmHg Melting Pt: –19 °C Density: 1.2 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00000521 CAS Number: 1461-22-9 EINECS: 215-958-7 UN: 2788 ADR: 6.1,II |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...