Methoxytrimethylsilane ≥97%
Supplier: BeanTown Chemical
Synonyms:
Trimethyl(methoxy)silane
143030-10G
143030-250G
143030-50G
BT143030-10GEA
23
USD
BT143030-10G
BT143030-250G
BT143030-50G
Methoxytrimethylsilane ≥97%
Methoxytrimethylsilane
CAS: 1825-61-2; EC No: 217-369-0; MDL No: MFCD00008344
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3OSI(CH3)3; MW: 104.22
Boiling Point: 57-58°; Flash point: -20°C (-4°F)
Density (g/mL): 0.756; Refractive Index: 1.366
Moisture Sensitive
- Refractive index: 1.366
- Moisture Sensitive
- Liquid
Formula:
C₄H₁₂OSi MW: 104.22 g/mol Boiling Pt: 57…58 °C Density: 0.756 g/mL Flash Pt: –20 °C (–4 °F) |
MDL Number:
MFCD00008344 CAS Number: 1825-61-2 EINECS: 217-369-0 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...