2,4-Dichlorobenzoyl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,4-Dichlorobenzene-1-carbonyl chloride

142995-250G 142995-50G
BT142995-250GEA 112 USD
BT142995-250G BT142995-50G
2,4-Dichlorobenzoyl chloride ≥98%
2,4-Dichlorobenzoyl chloride

CAS: 89-75-8; EC No: 201-936-4; MDL No: MFCD00000661; RTECS: DM6636766 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C7H3Cl3O ; MW: 209.46 Melting Point: 16-18°; Boiling Point: 150°/34 mmHg; Flash point: 137°C (238°F) Density (g/mL): 1.494; Refractive Index: 1.592 Moisture Sensitive


  • RTECS: DM6636766
  • Refractive index: 1.592
  • Moisture Sensitive
  • Liquid

Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 150 °C/34 mmHg
Melting Pt: 16…18 °C
Density: 1.494 g/mL
Flash Pt: 137 °C (238 °F)
MDL Number: MFCD00000661
CAS Number: 89-75-8
EINECS: 201-936-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR