3-Bromo-4-fluoroaniline ≥98%

Supplier: BeanTown Chemical

142880-1G 142880-25G 142880-5G
BT142880-1GEA 30 USD
BT142880-1G BT142880-25G BT142880-5G
3-Bromo-4-fluoroaniline ≥98%
3-Bromo-4-fluoroaniline

CAS: 656-64-4; MDL No: MFCD03788559 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C6H5BrFN; MW: 190.01 Melting Point: 34-37°


  • Solid

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Melting Pt: 34…37 °C
MDL Number: MFCD03788559
CAS Number: 656-64-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR