2-Amino-3-methylpyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Methyl-2-pyridylamine, 3-Methylpyridin-2-amine, 2-Amino-3-picoline

142855-100G 142855-25G 142855-500G
BT142855-100GEA 40 USD
BT142855-100G BT142855-25G BT142855-500G
2-Amino-3-methylpyridine ≥97%
2-Amino-3-methylpyridine

CAS: 1603-40-3; EC No: 216-501-4; MDL No: MFCD00006320; RTECS: US1850000 UN No: UN2811; Haz Class: 6.1; Packing Group: II Solid; Molecular Formula: C6H8N2 ; MW: 108.14 Melting Point: 29-31°; Boiling Point: 221-222°; Flash point: 118°C (244°F) Density (g/mL): 1.073; Refractive Index: 1.582 Hygroscopic


  • RTECS: US1850000
  • Refractive index: 1.582
  • Hygroscopic
  • Solid

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 221.1
Melting Pt: 29…31 °C
Density: 1.073 g/mL
Flash Pt: 118 °C (244 °F)
MDL Number: MFCD00006320
CAS Number: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR