Ammonium hydrogen sulphate ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Ammonium bisulphate
142650-100G
142650-25G
142650-500G
BT142650-100GEA
76
USD
BT142650-100G
BT142650-25G
BT142650-500G
Ammonium hydrogen sulphate ≥99.9%
Ammonium hydrogen sulphate
CAS: 7803-63-6; EC No: 232-265-5; MDL No: MFCD00051073
UN No: UN2506; Haz Class: 8; Packing Group: II
Lump; Linear Formula: NH4HSO4; MW: 115.11
Melting Point: 121-145°; Boiling Point: 350° (decomposes)
Density (g/mL): 1.79
Hygroscopic
- Hygroscopic
- Lump
Formula:
H₅NO₄S MW: 115.11 g/mol Boiling Pt: 350 (dec.) Melting Pt: 138.85 Density: 1.79 g/mL |
MDL Number:
MFCD00051073 CAS Number: 7803-63-6 EINECS: 232-265-5 UN: 2506 ADR: 8 II Merck Index: 12,00526 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...