Barium titanate ≥99%, high surface area
Supplier: BeanTown Chemical
Synonyms:
Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
142645-100G
142645-25G
BT142645-100GEA
203.35
USD
BT142645-100G
BT142645-25G
Barium titanate ≥99%, high surface area
Barium titanate
CAS: 12047-27-7; EC No: 234-975-0; MDL No: MFCD00003447
UN No: UN1564; Haz Class: 6.1; Packing Group: III
0.05-0.07 Micron APS Powder, S.A. 10-30 m2/g ; Molecular Formula: BaTiO3; MW: 233.19
Density (g/mL): 6.08
- 0.05-0.07 Micron APS Powder, S.A. 10-30 m²/g
Formula:
BaO₃Ti MW: 233.21 g/mol Melting Pt: 1625 Density: 6.08 g/mL |
MDL Number:
MFCD00003447 CAS Number: 12047-27-7 EINECS: 234-975-0 UN: 1564 ADR: 6.1 III Merck Index: 12,01029 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...