N-Ethylethylenediamine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Aminoethyl(ethyl)amine

142630-25G 142630-5G
BT142630-25GEA 88 USD
BT142630-25G BT142630-5G
N-Ethylethylenediamine ≥98%
N-Ethylethylenediamine

CAS: 110-72-5; EC No: 203-795-4; MDL No: MFCD00008166 UN No: UN2733; Haz Class: 3 (8); Packing Group: III Liquid; Linear Formula: CH3CH2NHCH2CH2NH2; Molecular Formula: C4H12N2; MW: 88.15 Boiling Point: 128-130°; Flash point: 10°C (50°F) Density (g/mL): 0.837 ; Refractive Index: 1.439 Air Sensitive


  • Refractive index: 1.439
  • Air Sensitive
  • Liquid

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 128…130 °C
Density: 0.837 g/mL
Flash Pt: 10 °C (50 °F)
MDL Number: MFCD00008166
CAS Number: 110-72-5
EINECS: 203-795-4
UN: 2733
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR