Tetramethylammonium hydroxide 1.0 M in water ACS
Supplier: BeanTown Chemical
142595-1L
142595-250ML
BT142595-1LEA
130
USD
BT142595-1L
BT142595-250ML
Tetramethylammonium hydroxide 1.0 M in water ACS
Tetramethylammonium hydroxide
CAS: 75-59-2; EC No: 200-882-9; MDL No: MFCD00008280
UN No: UN1835; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: (CH3)4NOH ; Molecular Formula: C4H13NO; MW: 91.15
Air Sensitive
- Air Sensitive
- Liquid
Formula:
C₄H₁₃NO |
MDL Number:
MFCD00008280 CAS Number: 75-59-2 EINECS: 200-882-9 UN: 1835 ADR: 8,II |
Specification Test Results
Assay | 1.0±0.02 mol/L |
Color (APHA) | ≤10 |
Residue after ignition | ≤5ppm |
Chloride (Cl) | ≤0.03% |
Potassium (K) | ≤2ppm |
Sodium (Na) | ≤2ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...