Magnesium nitrate hexahydrate ≥98%
Supplier: BeanTown Chemical
142570-1KG
142570-250G
142570-5KG
BT142570-1KGEA
63.66
USD
BT142570-1KG
BT142570-250G
BT142570-5KG
Magnesium nitrate hexahydrate ≥98%
Magnesium nitrate hexahydrate
CAS: 13446-18-9; EC No: 233-826-7; MDL No: MFCD00149779; RTECS: OM3756000
UN No: UN1474; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Mg(NO3)2·6H2O ; MW: 256.41
Melting Point: 89° (decomposes); Boiling Point: 330°
Density (g/mL): 1.636
Hygroscopic
- RTECS: OM3756000
- Hygroscopic
- Crystalline
Formula:
Mg(NO₃)₂·6H₂O MW: 256.41 g/mol Boiling Pt: 330 °C Melting Pt: 89 °C (dec.) Density: 1.636 g/mL |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 EINECS: 233-826-7 UN: 1474 ADR: 5.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...