Silver metavanadate

Supplier: BeanTown Chemical

142495-25G 142495-5G
BT142495-25GEA 240 USD
BT142495-25G BT142495-5G
Silver metavanadate
Silver metavanadate

CAS: 13497-94-4; EC No: 236-820-2; MDL No: MFCD00041760 UN No: UN3285; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: AgO3V; MW: 206.81 Light Sensitive


  • Light Sensitive
  • Powder

Formula: AgO₃V
MW: 206.807 g/mol
MDL Number: MFCD00041760
CAS Number: 13497-94-4
EINECS: 236-820-2
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR