(R)-(-)-Epichlorhydrin ≥98%
Supplier: BeanTown Chemical
Synonyms:
(R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane
142475-1G
142475-25G
142475-5G
BT142475-1GEA
52
USD
BT142475-1G
BT142475-25G
BT142475-5G
(R)-(-)-Epichlorhydrin ≥98%
(R)-(-)-Epichlorhydrin
CAS: 51594-55-9; EC No: 424-280-2; MDL No: MFCD00077759; RTECS: RR0427000
UN No: UN2023; Haz Class: 6.1 (3); Packing Group: II
Liquid; Molecular Formula: C3H5ClO; MW: 92.52
Flash point: 32°C (90°F)
Density (g/mL): 1.18; Optical Rotation: [α]20/D −34°, c = 1 in methanol
- RTECS: RR0427000
- Optical rotation: [α]20/D −34°, c = 1 in methanol
- Liquid
Formula:
ClCH₂CH(O)CH₂ MW: 92.52 g/mol Density: 1.18 g/mL Flash Pt: 32 °C (90 °F) |
MDL Number:
MFCD00077759 CAS Number: 51594-55-9 EINECS: 424-280-2 UN: 2023 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...