Nitrosium hexafluorophosphate ≥96%
Supplier: BeanTown Chemical
Synonyms:
Nitrosonium hexafluorophosphate, Nitrosyl hexafluorophosphate
142415-25G
142415-5G
BT142415-25GEA
416
USD
BT142415-25G
BT142415-5G
Nitrosium hexafluorophosphate ≥96%
Nitrosium hexafluorophosphate
CAS: 16921-91-8; EC No: 240-980-9; MDL No: MFCD00040324
UN No: UN3260; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: PF6NO; MW: 174.97
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
F₆NOP MW: 174.97 g/mol |
MDL Number:
MFCD00040324 CAS Number: 16921-91-8 EINECS: 240-980-9 UN: 3260 ADR: 8,II |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...