Sodium hexafluoroaluminate ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Trisodium hexafluoroaluminate, Sodium hexafluoroaluminate(III)

142375-250G 142375-50G
BT142375-250GEA 407 USD
BT142375-250G BT142375-50G
Sodium hexafluoroaluminate ≥99.5% (trace metals basis)
Sodium hexafluoroaluminate

CAS: 13775-53-6; EC No: 237-410-6; MDL No: MFCD00003507; RTECS: BD0075000 UN No: UN3260; Haz Class: 8; Packing Group: III Lump; Linear Formula: Na3AlF6 ; MW: 209.95 Melting Point: 1000° Density (g/mL): 2.90


  • RTECS: BD0075000
  • Lump

Formula: AlF₆Na₃
MW: 209.94 g/mol
Melting Pt: 1000 °C
Density: 2.90 g/mL
MDL Number: MFCD00003507
CAS Number: 13775-53-6
EINECS: 237-410-6
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR