Decahydronaphthalene (mixture of cis and trans isomers) ≥98%
Supplier: BeanTown Chemical
Synonyms:
Decalin, Bicyclo[4.4.0]decane, Decahydronaphthalene
142330-100ML
142330-2.5L
142330-500ML
BT142330-100MLEA
26.98
USD
BT142330-100ML
BT142330-2.5L
BT142330-500ML
Decahydronaphthalene (mixture of cis and trans isomers) ≥98%
Decahydronaphthalene (mixture of cis and trans isomers)
CAS: 91-17-8; EC No: 202-046-9; MDL No: MFCD00004130; RTECS: QJ3150000
UN No: UN1147; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C10H18; MW: 138.25
Melting Point: -125°; Boiling Point: 189-191°; Flash point: 57°C (135°F)
Density (g/mL): 0.896; Refractive Index: 1.474
- RTECS: QJ3150000
- Refractive index: 1.474
- Liquid
Formula:
C₁₀H₁₈ MW: 138.25 g/mol Boiling Pt: 189…191 °C Melting Pt: –125 °C Density: 0.896 g/mL Flash Pt: 57 °C (135 °F) |
MDL Number:
MFCD00004130 CAS Number: 91-17-8 EINECS: 202-046-9 UN: 1147 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...