Nickel(II) sulphate hexahydrate ≥99.95%
Supplier: BeanTown Chemical
Synonyms:
Nickelous sulphate, Hexahydrate, Nickel sulphate hexahydrate
142325-250G
142325-50G
BT142325-250GEA
297
USD
BT142325-250G
BT142325-50G
Nickel(II) sulphate hexahydrate ≥99.95%
Nickel(II) sulphate hexahydrate
CAS: 10101-97-0; EC No: 232-104-9; MDL No: MFCD00149813; RTECS: QR9600000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Linear Formula: NiSO4·6H2O; MW: 262.86
Density (g/mL): 2.070
- RTECS: QR9600000
- Crystalline/Powder
Formula:
NiSO₄·6H₂O MW: 262.85 g/mol Melting Pt: 840 Density: 2.070 g/mL |
MDL Number:
MFCD00149813 CAS Number: 10101-97-0 EINECS: 232-104-9 UN: 3288 ADR: 6.1 III Merck Index: 13,06541 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...