2,4,4-Trimethyl-1-pentene ≥99%
Supplier: BeanTown Chemical
142270-100G
142270-25G
BT142270-100GEA
128
USD
BT142270-100G
BT142270-25G
2,4,4-Trimethyl-1-pentene ≥99%
2,4,4-Trimethyl-1-pentene
CAS: 107-39-1; EC No: 203-486-4; MDL No: MFCD00008855; RTECS: SB2717300
UN No: UN2050; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)3CCH2C(CH3)=CH2; Molecular Formula: C8H16; MW: 112.21
Boiling Point: 101-102°; Flash point: -3°C (27°F)
Density (g/mL): 0.708; Refractive Index: 1.408
- RTECS: SB2717300
- Refractive index: 1.408
- Liquid
Formula:
C₈H₁₆ Boiling Pt: 101…102 °C Density: 0.708 g/mL Flash Pt: –3 °C (27 °F) |
MDL Number:
MFCD00008855 CAS Number: 107-39-1 EINECS: 203-486-4 UN: 2050 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...