4-Amino-1-boc-piperidine ≥97%

Supplier: BeanTown Chemical

Synonyms: tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester

142205-1G 142205-5G
BT142205-1GEA 68 USD
BT142205-1G BT142205-5G
4-Amino-1-boc-piperidine ≥97%
4-Amino-1-Boc-piperidine

CAS: 87120-72-7; MDL No: MFCD01076201 UN No: UN3259; Haz Class: 8; Packing Group: III Solid; Molecular Formula: C10H20N2O2; MW: 200.28 Melting Point: 44-48° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₁₀H₂₀N₂O₂
Melting Pt: 44…48 °C
MDL Number: MFCD01076201
CAS Number: 87120-72-7
ADR: 8,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR