Dimethylketene methyl trimethylsilyl acetal ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, ((1-Methoxy-2-methylprop-1-en-1-yl)oxy)trimethylsilane

142120-25G 142120-5G
BT142120-25GEA 246 USD
BT142120-25G BT142120-5G
Dimethylketene methyl trimethylsilyl acetal ≥97%
Dimethylketene methyl trimethylsilyl acetal

CAS: 31469-15-5; MDL No: MFCD00010232 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C8H18O2Si; MW: 174.31 Boiling Point: 35°/15 mmHg; Flash point: 30°C (86°F) Density (g/mL): 0.858; Refractive Index: 1.415 Moisture Sensitive


  • Refractive index: 1.415
  • Moisture Sensitive
  • Liquid

Formula: C₈H₁₈O₂Si
MW: 174.32 g/mol
Boiling Pt: 35 °C/15 mmHg
Density: 0.858 g/mL
Flash Pt: 30 °C (86 °F)
MDL Number: MFCD00010232
CAS Number: 31469-15-5
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR