1-Bromo-2-methoxyethane ≥96% stabilized

Supplier: BeanTown Chemical

Synonyms: 2-Bromoethyl methyl ether

142090-100G 142090-25G 142090-5G
BT142090-100GEA 174 USD
BT142090-100G BT142090-25G BT142090-5G
1-Bromo-2-methoxyethane ≥96% stabilized
1-Bromo-2-methoxyethane

CAS: 6482-24-2; EC No: 229-339-4; MDL No: MFCD00000236 UN No: UN3271; Haz Class: 3; Packing Group: III Liquid; Linear Formula: BrCH2CH2OCH3; Molecular Formula: C3H7BrO; MW: 138.99 Boiling Point: 40-41°/66 mmHg; Flash point: 28°C (82°F) Density (g/mL): 1.479; Refractive Index: 1.447


  • Refractive index: 1.447
  • Liquid

Formula: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Boiling Pt: 40…41 °C/66 mmHg
Density: 1.479 g/mL
Flash Pt: 28 °C (82 °F)
MDL Number: MFCD00000236
CAS Number: 6482-24-2
EINECS: 229-339-4
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR