Mercaptoacetic acid (thioglycolic acid) ≥97%

Supplier: BeanTown Chemical

Synonyms: Mercaptoacetic acid

142035-100G 142035-2.5KG 142035-500G
BT142035-100GEA 30.99 USD
BT142035-100G BT142035-2.5KG BT142035-500G
Mercaptoacetic acid (thioglycolic acid) ≥97%
Mercaptoacetic acid (thioglycolic acid)

CAS: 68-11-1; EC No: 200-677-4; MDL No: MFCD00004876; RTECS: AI5950000 UN No: UN1940; Haz Class: 8; Packing Group: II Liquid; Linear Formula: HSCH2COOH; Molecular Formula: C2H4O2S; MW: 92.11 Melting Point: -16°; Boiling Point: 220°; Flash point: 119°C (246°F) Density (g/mL): 1.326; Refractive Index: 1.505 Air Sensitive


  • RTECS: AI5950000
  • Refractive index: 1.505
  • Air Sensitive
  • Liquid

Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 96
Melting Pt: -16.5
Density: 1.326 g/mL
Flash Pt: 119 °C (246 °F)
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: 8 II
Merck Index: 13,09410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45179825145ec44f2e058197a0ee54845ae7c750&vid=51e43f5b69b880b196570b3065f9b4d8db7336d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR