Neodymium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Supplier: BeanTown Chemical
142005-25X25MM
142005-50X50MM
BT142005-25X25EA
131
USD
BT142005-25X25
BT142005-50X50
Neodymium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Neodymium
CAS: 7440-00-8; EC No: 231-109-3; MDL No: MFCD00011130
UN No: UN3178; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: Nd; MW: 144.24
Melting Point: 1021°; Boiling Point: 3074°
Density (g/mL): 7.003
- Solid
Formula:
Nd MW: 144.24 g/mol Boiling Pt: 3074 °C Melting Pt: 1021 °C Density: 7.003 g/mL |
MDL Number:
MFCD00011130 CAS Number: 7440-00-8 EINECS: 231-109-3 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...