Bismuth(III) nitrate pentahydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Bismuth trinitrate pentahydrate

141995-100G 141995-500G
BT141995-100GEA 43.12 USD
BT141995-100G BT141995-500G
Bismuth(III) nitrate pentahydrate ≥98%
Bismuth(III) nitrate pentahydrate

CAS: 10035-06-0; EC No: 233-791-8; MDL No: MFCD00149157 UN No: UN1477; Haz Class: 5.1; Packing Group: II Solid; Linear Formula: Bi(NO3)3·5H2O; MW: 485.07 Melting Point: 30°; Boiling Point: 75-80° Hygroscopic


  • Hygroscopic
  • Solid

Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 75…80 °C
Melting Pt: 30 °C
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR