Iron(III) fluoride ≥97%

Supplier: BeanTown Chemical

Synonyms: Iron trifluoride, Ferric fluoride

141980-10G 141980-50G
BT141980-10GEA 32 USD
BT141980-10G BT141980-50G
Iron(III) fluoride ≥97%
Iron(III) fluoride

CAS: 7783-50-8; EC No: 232-002-4; MDL No: MFCD00011012 UN No: UN3260; Haz Class: 8; Packing Group: II Powder; Linear Formula: FeF3; MW: 112.84 Hygroscopic


  • Hygroscopic
  • Powder

Formula: F₃Fe
MW: 112.84 g/mol
MDL Number: MFCD00011012
CAS Number: 7783-50-8
EINECS: 232-002-4
UN: 3260
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR