4-Fluorobenzonitrile ≥99%
Supplier: BeanTown Chemical
Synonyms:
p-Fluorobenzonitrile
141845-100G
141845-25G
141845-5G
BT141845-100GEA
251
USD
BT141845-100G
BT141845-25G
BT141845-5G
4-Fluorobenzonitrile ≥99%
4-Fluorobenzonitrile
CAS: 1194-02-1; EC No: 214-784-9; MDL No: MFCD00001812; RTECS: DI4368500
UN No: UN1325; Haz Class: 4.1; Packing Group: II
Crystalline; Molecular Formula: C7H4FN; MW: 121.11
Melting Point: 32-34°; Boiling Point: 188°/750 mmHg; Flash point: 65°C (149°F)
- RTECS: DI4368500
- Crystalline
Formula:
C₇H₄FN MW: 121.11 g/mol Boiling Pt: 188 °C/750 mmHg Melting Pt: 32…34 °C Flash Pt: 65 °C (149 °F) |
MDL Number:
MFCD00001812 CAS Number: 1194-02-1 EINECS: 214-784-9 UN: 1325 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...