2-Chloro-p-toluidine ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Chloro-4-methylaniline, 4-Amino-3-chlorotoluene
141815-100G
141815-25G
141815-5G
BT141815-100GEA
341
USD
BT141815-100G
BT141815-25G
BT141815-5G
2-Chloro-p-toluidine ≥98%
2-Chloro-p-toluidine
CAS: 615-65-6; EC No: 210-440-7; MDL No: MFCD00007666; RTECS: XU5110000
UN No: UN3429; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C7H8ClN; MW: 141.60
Melting Point: 7°; Flash point: 99°C (210°F)
Density (g/mL): 1.151 ; Refractive Index: 1.575
- RTECS: XU5110000
- Refractive index: 1.575
- Liquid
Formula:
C₇H₈ClN MW: 141.6 g/mol Melting Pt: 7 °C Density: 1.151 g/mL Flash Pt: 99 °C (210 °F) |
MDL Number:
MFCD00007666 CAS Number: 615-65-6 EINECS: 210-440-7 UN: 3429 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...