2-Methyl-1-pyrroline

Supplier: BeanTown Chemical

141745-25ML 141745-5ML
BT141745-25MLEA 200 USD
BT141745-25ML BT141745-5ML
2-Methyl-1-pyrroline
2-Methyl-1-pyrroline

CAS: 872-32-2; EC No: 212-822-9; MDL No: MFCD00128806 UN No: UN2733; Haz Class: 3 (8); Packing Group: II Liquid; Molecular Formula: C5H9N; MW: 83.13 Boiling Point: 104-105°; Flash point: 11°C (52°F) Density (g/mL): 0.878; Refractive Index: 1.444


  • Refractive index: 1.444
  • Liquid

Formula: C₅H₉N
MW: 83.13 g/mol
Boiling Pt: 104…105 °C
Density: 0.878 g/mL
Flash Pt: 11 °C (52 °F)
MDL Number: MFCD00128806
CAS Number: 872-32-2
EINECS: 212-822-9
ADR: 3 (8),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR