Diiodomethane ≥99% stabilized

Supplier: BeanTown Chemical

Synonyms: Methylene iodide

141645-100G 141645-25G
BT141645-100GEA 119.59 USD
BT141645-100G BT141645-25G
Diiodomethane ≥99% stabilized
Diiodomethane

CAS: 75-11-6; EC No: 200-841-5; MDL No: MFCD00001079; RTECS: PA8575000 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: CH2I2; MW: 267.84 Melting Point: 5-8°; Boiling Point: 67-69°/11 mmHg; Flash point: 110°C (230°F) Density (g/mL): 3.325; Refractive Index: 1.740 Light Sensitive


  • RTECS: PA8575000
  • Refractive index: 1.740
  • Light Sensitive
  • Liquid

Formula: CH₂I₂
MW: 267.84 g/mol
Boiling Pt: 67…69 °C/11 mmHg
Melting Pt: 5…8 °C
Density: 3.325 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00001079
CAS Number: 75-11-6
EINECS: 200-841-5
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e37a85a8930c2cbeaf62626d427944a69722cc72&vid=73a1847aa5acf8fa3b0e777fe1bcae9e4f7bde3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR