Tributylphosphine ≥94%
Supplier: BeanTown Chemical
Synonyms:
Tri-n-butylphosphine, TBP
141635-250G
141635-50G
BT141635-250GEA
112
USD
BT141635-250G
BT141635-50G
Tributylphosphine ≥94%
Tributylphosphine
CAS: 998-40-3; EC No: 213-651-2; MDL No: MFCD00009462; RTECS: SZ3270000
UN No: UN3254; Haz Class: 4.2; Packing Group: I
Liquid; Linear Formula: [CH3(CH2)3]3P; Molecular Formula: C12H27P; MW: 202.32
Melting Point: -60°; Boiling Point: 150°/50 mmHg; Flash point: 37°C (99°F)
Density (g/mL): 0.810; Refractive Index: 1.462
Air Sensitive, Moisture Sensitive
- RTECS: SZ3270000
- Refractive index: 1.462
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₁₂H₂₇P MW: 202.32 g/mol Boiling Pt: 150 °C/50 mmHg Melting Pt: –60 °C Density: 0.810 g/mL Flash Pt: 37 °C (99 °F) |
MDL Number:
MFCD00009462 CAS Number: 998-40-3 EINECS: 213-651-2 UN: 3254 ADR: 4.2,I |
Specification Test Results
Assay | 94% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...