Potassium hydrogen difluoride ≥98%

Supplier: BeanTown Chemical

141605-100G 141605-2.5KG 141605-500G
BT141605-100GEA 25.09 USD
BT141605-100G BT141605-2.5KG BT141605-500G
Potassium hydrogen difluoride ≥98%
Potassium hydrogen difluoride

CAS: 7789-29-9; EC No: 232-156-2; MDL No: MFCD00011403; RTECS: TS6650000 UN No: UN1811; Haz Class: 8 (6.1); Packing Group: II Crystalline; Linear Formula: KHF2; MW: 78.10 Melting Point: 239° Density (g/mL): 2.37 Hygroscopic


  • RTECS: TS6650000
  • Hygroscopic
  • Crystalline

Formula: HF₂K
Melting Pt: 239 °C
Density: 2.37 g/mL
MDL Number: MFCD00011403
CAS Number: 7789-29-9
EINECS: 232-156-2
UN: 1811
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR