4-Fluorobenzaldehyde ≥98%
Supplier: BeanTown Chemical
141515-100G
141515-25G
141515-500G
BT141515-100GEA
92
USD
BT141515-100G
BT141515-25G
BT141515-500G
4-Fluorobenzaldehyde ≥98%
4-Fluorobenzaldehyde
CAS: 459-57-4; EC No: 207-293-6; MDL No: MFCD00003378
UN No: UN1989; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C7H5FO; MW: 124.11
Melting Point: -10°; Boiling Point: 181°; Flash point: 56°C (133°F)
Density (g/mL): 1.157; Refractive Index: 1.521
Air Sensitive
- Refractive index: 1.521
- Air Sensitive
- Liquid
Formula:
C₇H₅FO MW: 124.11 g/mol Boiling Pt: 181 °C Melting Pt: –10 °C Density: 1.157 g/mL Flash Pt: 56 °C (133 °F) |
MDL Number:
MFCD00003378 CAS Number: 459-57-4 EINECS: 207-293-6 UN: 1989 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...