(±)-1-Methoxy-2-propanol ≥99%

Supplier: BeanTown Chemical

Synonyms: (±)-Monopropylene glycol methyl ether, 1-Methoxy-2-propanol

141490-1L 141490-250ML 141490-4L
BT141490-1LEA 40.56 USD
BT141490-1L BT141490-250ML BT141490-4L
(±)-1-Methoxy-2-propanol ≥99%
(±)-1-Methoxy-2-propanol

CAS: 107-98-2; EC No: 203-539-1; MDL No: MFCD00004537; RTECS: UB7700000 UN No: UN3092 ; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C4H10O2; MW: 90.12 Boiling Point: 118-119°; Flash point: 34°C (93°F) Density (g/mL): 0.922; Refractive Index: 1.403


  • RTECS: UB7700000
  • Refractive index: 1.403
  • Liquid

Formula: CH₃CH(OH)CH₂OCH₃
MW: 90.12 g/mol
Boiling Pt: 118…119 °C
Density: 0.922 g/mL
Flash Pt: 34 °C (93 °F)
MDL Number: MFCD00004537
CAS Number: 107-98-2
EINECS: 203-539-1
UN: 3092
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e076d8e7a4ef95472e37d2c355f37d64eac8ee&vid=26da88ae9f09b8195abc9bdd949f6bbdaf429255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR