2,4-Dichloro-3-cyanopyridine ≥95%

Supplier: BeanTown Chemical

Synonyms: 2,4-Dichloropyridine-3-carbonitrile, 2,4-Dichloronicotinonitrile

141355-1G 141355-250MG 141355-5G
BT141355-1GEA 88 USD
BT141355-1G BT141355-250MG BT141355-5G
2,4-Dichloro-3-cyanopyridine ≥95%
2,4-Dichloro-3-cyanopyridine

CAS: 180995-12-4 UN No: UN3439; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C6H2Cl2N2; MW: 173.00


  • Solid

Formula: C₆H₂Cl₂N₂
MW: 172.996 g/mol
MDL Number: MFCD09750302
CAS Number: 180995-12-4
ADR: 6.1,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR