2-Aminothiophenol ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Aminophenyl mercaptan, 2-Aminobenzenethiol, 2-Mercaptoaniline

141325-100G 141325-25G
BT141325-100GEA 63 USD
BT141325-100G BT141325-25G
2-Aminothiophenol ≥98%
2-Aminothiophenol

CAS: 137-07-5; EC No: 205-277-3; MDL No: MFCD00007702; RTECS: DC0600000 UN No: UN3267; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C6H7NS; MW: 125.19 Melting Point: 16-20°; Boiling Point: 70-72°/0.2 mmHg; Flash point: 79°C (174°F) Density (g/mL): 1.17; Refractive Index: 1.642 Air Sensitive


  • RTECS: DC0600000
  • Refractive index: 1.642
  • Air Sensitive
  • Liquid

Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 70…72 °C/0.2 mmHg
Melting Pt: 16…20 °C
Density: 1.17 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00007702
CAS Number: 137-07-5
EINECS: 205-277-3
UN: 3267
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR