1,10-Phenanthroline monohydrate ACS
Supplier: BeanTown Chemical
Synonyms:
1,10-Phenanthroline hydrate
141250-10G
141250-50G
BT141250-10GEA
66.82
USD
BT141250-10G
BT141250-50G
1,10-Phenanthroline monohydrate ACS
1,10-Phenanthroline monohydrate
CAS: 5144-89-8; EC No: 200-629-2; MDL No: MFCD00011678; RTECS: SF8437000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C12H8N2·H2O ; MW: 198.22
Melting Point: 100-104°
- RTECS: SF8437000
- Crystalline
Formula:
C₁₂H₈N₂·1H₂O Melting Pt: 100…104 °C |
MDL Number:
MFCD00149973 CAS Number: 5144-89-8 EINECS: 200-629-2 UN: 2811 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...