1-Propanol, anhydrous ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

141210-1L 141210-250ML 141210-4L
BT141210-1LEA 89.59 USD
BT141210-1L BT141210-250ML BT141210-4L
1-Propanol, anhydrous ≥99.9%
1-Propanol

CAS: 71-23-8; EC No: 200-746-9; MDL No: MFCD00002941; RTECS: UH8225000 UN No: UN1274; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH2CH2OH; Molecular Formula: C3H8; MW: 60.10 Melting Point: -127°; Boiling Point: 97°; Flash point: 22°C (72°F) Density (g/mL): 0.804; Refractive Index: 1.384


  • RTECS: UH8225000
  • Refractive index: 1.384
  • Liquid

Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97 °C
Melting Pt: –127 °C
Density: 0.804 g/mL
Flash Pt: 22 °C (72 °F)
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR