2-Fluorophenol ≥98%

Supplier: BeanTown Chemical

Synonyms: o-Fluorophenol

141170-100G 141170-25G 141170-500G
BT141170-100GEA 176 USD
BT141170-100G BT141170-25G BT141170-500G
2-Fluorophenol ≥98%
2-Fluorophenol

CAS: 367-12-4; EC No: 206-681-2; MDL No: MFCD00002155; RTECS: SL4530000 UN No: UN1993 ; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C6H5FO; MW: 112.10 Melting Point: 16.1°; Boiling Point: 171-172°/741 mmHg; Flash point: 47°C (117°F) Density (g/mL): 1.256; Refractive Index: 1.511


  • RTECS: SL4530000
  • Refractive index: 1.511
  • Liquid

Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 171…172 °C/741 mmHg
Melting Pt: 16.1 °C
Density: 1.256 g/mL
Flash Pt: 47 °C (117 °F)
MDL Number: MFCD00002155
CAS Number: 367-12-4
EINECS: 206-681-2
UN: 1993
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR