Ethyl trifluoroacetate ≥99%

Supplier: BeanTown Chemical

Synonyms: Trifluoroacetic ethyl ester

141160-1KG 141160-250G 141160-50G
BT141160-1KGEA 719 USD
BT141160-1KG BT141160-250G BT141160-50G
Ethyl trifluoroacetate ≥99%
Ethyl trifluoroacetate

CAS: 383-63-1; EC No: 206-851-6; MDL No: MFCD00000419 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: CF3COOCH2CH3; Molecular Formula: C4H5F3O2; MW: 142.08 Boiling Point: 60-62°; Flash point: -1°C (30°F) Density (g/mL): 1.194; Refractive Index: 1.307 Moisture Sensitive


  • Refractive index: 1.307
  • Moisture Sensitive
  • Liquid

Formula: C₄H₅F₃O₂
Boiling Pt: 60…62 °C
Density: 1.194 g/mL
Flash Pt: –1 °C (30 °F)
MDL Number: MFCD00000419
CAS Number: 383-63-1
EINECS: 206-851-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR