Silver(II) oxide ≥98%

Supplier: BeanTown Chemical

141155-25G 141155-5G
BT141155-25GEA 158.63 USD
BT141155-25G BT141155-5G
Silver(II) oxide ≥98%
Silver(II) oxide

CAS: 1301-96-8; EC No: 215-098-2; MDL No: MFCD00044285 UN No: UN3085; Haz Class: 5.1 (8); Packing Group: II Powder; Molecular Formula: AgO; MW: 123.87 Melting Point: 100° (decomposes) Density (g/mL): 7.483 Light Sensitive, Moisture Sensitive


  • Light Sensitive, Moisture Sensitive
  • Powder

Formula: AgO
MW: 123.87 g/mol
Melting Pt: >100 °C (dec.)
Density: 7.483 g/mL
MDL Number: MFCD00044285
CAS Number: 1301-96-8
EINECS: 215-098-2
UN: 3085
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR